accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 88738-86-7

IUPAC Name: methyl 3-[4-(bromomethyl)phenyl]prop-2-enoate
Synonyms: methyl 3-[4-(bromomethyl)phenyl]prop-2-enoate
Molecular Weight (g/mol): 255.11
Molecular Formula: C11H11BrO2
InChi Key: ZSRCGGBALFGALF-UHFFFAOYSA-N
SMILES: COC(=O)C=CC1=CC=C(CBr)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results