accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 88740-77-6

IUPAC Name: 3-(2-chloro-6-fluorophenyl)propanoic acid
Synonyms: 3-(2-chloro-6-fluorophenyl)propanoic acid
Molecular Weight (g/mol): 202.61
Molecular Formula: C9H8ClFO2
InChi Key: IJIZOJZUZZTIDJ-UHFFFAOYSA-N
SMILES: OC(=O)CCC1=C(F)C=CC=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results