accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 888041-37-0

IUPAC Name: 2,7-dibromotriphenylene
Synonyms: 2,7-dibromotriphenylene
Molecular Weight (g/mol): 386.09
Molecular Formula: C18H10Br2
InChi Key: BPGPBYGXGRDFQG-UHFFFAOYSA-N
SMILES: BrC1=CC=C2C3=CC=C(Br)C=C3C3=CC=CC=C3C2=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results