accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 88840-42-0

IUPAC Name: 1-methyl-1-propylpiperidin-1-ium bromide
Synonyms: 1-methyl-1-propylpiperidin-1-ium bromide
Molecular Weight (g/mol): 222.17
Molecular Formula: C9H20BrN
InChi Key: CSANMXKHSHGXNC-UHFFFAOYSA-M
SMILES: [Br-].CCC[N+]1(C)CCCCC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results