accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 888504-28-7

IUPAC Name: potassium 5-methyl-1,3,4-oxadiazole-2-carboxylate
Synonyms: potassium 5-methyl-1,3,4-oxadiazole-2-carboxylate
Molecular Weight (g/mol): 166.18
Molecular Formula: C4H3KN2O3
InChi Key: BRHMYHXNUGVBCU-UHFFFAOYSA-M
SMILES: [K+].CC1=NN=C(O1)C([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results