accessibility menu, dialog, popup

UserName

Viewing profile as user:
BBrFFFK

CAS RN 888711-44-2

IUPAC Name: potassium (bromomethyl)trifluoroboranuide
Synonyms: potassium (bromomethyl)trifluoroboranuide
Molecular Weight (g/mol): 200.83
Molecular Formula: CH2BBrF3K
InChi Key: AZDFPIRYUOCVCJ-UHFFFAOYSA-N
SMILES: [K+].F[B-](F)(F)CBr
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)