accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 888711-47-5

IUPAC Name: potassium trifluoro(iodomethyl)boranuide
Synonyms: potassium trifluoro(iodomethyl)boranuide
Molecular Weight (g/mol): 247.83
Molecular Formula: CH2BF3IK
InChi Key: WQJVWRYIXQXISQ-UHFFFAOYSA-N
SMILES: [K+].F[B-](F)(F)CI
Molecular Weight (g/mol) 
  • (4)
  • (1)
Quantity 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (4)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)