accessibility menu, dialog, popup

UserName

Viewing profile as user:
BNFFFK

CAS RN 888711-58-8

IUPAC Name: potassium (cyanomethyl)trifluoroboranuide
Synonyms: potassium (cyanomethyl)trifluoroboranuide
Molecular Weight (g/mol): 146.95
Molecular Formula: C2H2BF3KN
InChi Key: IJENQMHWPRCCBA-UHFFFAOYSA-N
SMILES: [K+].F[B-](F)(F)CC#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)