accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CO(S)H3C(S)CH3(S)OH(R)CH3CH3OH3COO(S)H3C(R)OH(S)H3C(R)HO(R)OOONHOHO(S)CH3(R)OH3CCH3(Z)(Z)(Z)(E)(E)

CAS RN 88899-56-3

IUPAC Name: (2R,4R,5S,6R)-2-hydroxy-2-[(2S,3R,4S)-3-hydroxy-4-[(3S,4Z,6Z,9S,10S,11R,12Z,14E)-10-hydroxy-3,15-dimethoxy-7,9,11,13-tetramethyl-16-oxo-1-oxacyclohexadeca-4,6,12,14-tetraen-2-yl]pentan-2-yl]-5-methyl-6-(propan-2-yl)oxan-4-yl (2E)-3-[(2-hydroxy-5-oxocyclopent-1-en-1-yl)carbamoyl]prop-2-enoate
Synonyms: (2R,4R,5S,6R)-2-hydroxy-2-[(2S,3R,4S)-3-hydroxy-4-[(3S,4Z,6Z,9S,10S,11R,12Z,14E)-10-hydroxy-3,15-dimethoxy-7,9,11,13-tetramethyl-16-oxo-1-oxacyclohexadeca-4,6,12,14-tetraen-2-yl]pentan-2-yl]-6-isopropyl-5-methyloxan-4-yl (2E)-3-[(2-hydroxy-5-oxocyclopent-1-en-1-yl)carbamoyl]prop-2-enoate
Molecular Weight (g/mol): 816
Molecular Formula: C44H65NO13
InChi Key: KFUFLYSBMNNJTF-MZYUNMTESA-N
SMILES: CO[C@H]1\C=C/C=C(C)\C[C@H](C)[C@H](O)[C@H](C)\C=C(\C)/C=C(OC)\C(=O)OC1[C@@H](C)[C@@H](O)[C@H](C)[C@@]1(O)C[C@@H](OC(=O)\C=C\C(=O)NC2=C(O)CCC2=O)[C@H](C)[C@H](O1)C(C)C
Quantity 
  • (1)
Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results