accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 88912-24-7

IUPAC Name: 5,6-dichloropyridine-2-carboxylic acid
Synonyms: 5,6-dichloropyridine-2-carboxylic acid
Molecular Weight (g/mol): 192
Molecular Formula: C6H3Cl2NO2
InChi Key: QOJNAEPFSUYAFL-UHFFFAOYSA-N
SMILES: OC(=O)C1=NC(Cl)=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results