accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOOHCl

CAS RN 88912-27-0

IUPAC Name: 3-chloropyridine-4-carboxylic acid
Synonyms: 3-chloropyridine-4-carboxylic acid
Molecular Weight (g/mol): 157.55
Molecular Formula: C6H4ClNO2
InChi Key: MYAZXWFEMDJTFE-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(Cl)C=NC=C1
Grade 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results