accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOH3COCH3O

CAS RN 88917-22-0

IUPAC Name: 1-(3-methoxypropoxy)propyl acetate
Synonyms: ppg-2 methyl ether acetate
Molecular Weight (g/mol): 190.24
Molecular Formula: C9H18O4
InChi Key: USDXBNSCKZJDLC-UHFFFAOYNA-N
SMILES: CCC(OCCCOC)OC(C)=O
Boiling Point 
  • (1)
  • (2)
  • (4)
Percent Purity 
  • (1)
  • (4)
  • (2)
Quantity 
  • (3)
  • (1)
  • (2)
  • (1)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (3)
  • (4)
  • (1)
  • (2)
  • (4)
  • (1)
  • (4)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (4)
  • (4)