accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 889453-84-3

IUPAC Name: 2-[6-(4-methoxyphenoxy)-2-phenyl-8-(prop-2-en-1-yloxy)-hexahydro-2H-pyrano[3,2-d][1,3]dioxin-7-yl]-2,3-dihydro-1H-isoindole-1,3-dione
Synonyms: 2-[6-(4-methoxyphenoxy)-2-phenyl-8-(prop-2-en-1-yloxy)-hexahydro-2H-pyrano[3,2-d][1,3]dioxin-7-yl]isoindole-1,3-dione
Molecular Weight (g/mol): 543.57
Molecular Formula: C31H29NO8
InChi Key: KDSWOYVINHSEOI-UHFFFAOYNA-N
SMILES: COC1=CC=C(OC2OC3COC(OC3C(OCC=C)C2N2C(=O)C3=CC=CC=C3C2=O)C2=CC=CC=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results