accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 889672-73-5

CH3SnH3CCH3NSCl

CAS RN 889672-73-5

IUPAC Name: 2-chloro-5-(tributylstannyl)-1,3-thiazole
Synonyms: 2-chloro-5-(tributylstannyl)-1,3-thiazole
Molecular Weight (g/mol): 408.62
Molecular Formula: C15H28ClNSSn
InChi Key: ZXCCAZVZCUGOQU-UHFFFAOYSA-N
SMILES: CCCC[Sn](CCCC)(CCCC)C1=CN=C(Cl)S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results