accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 89-21-4

IUPAC Name: 4-chloro-1-methoxy-2-nitrobenzene
Synonyms: 4-chloro-1-methoxy-2-nitrobenzene
Molecular Weight (g/mol): 187.58
Molecular Formula: C7H6ClNO3
InChi Key: OSAYFGJUEOYRHY-UHFFFAOYSA-N
SMILES: COC1=CC=C(Cl)C=C1[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results