accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNOSH2NOO

CAS RN 89-29-2

IUPAC Name: 3-(3-methyl-5-oxo-4,5-dihydro-1H-pyrazol-1-yl)benzene-1-sulfonamide
Synonyms: 3-(3-methyl-5-oxo-4H-pyrazol-1-yl)benzenesulfonamide
Molecular Weight (g/mol): 253.28
Molecular Formula: C10H11N3O3S
InChi Key: ASVVGQURNHNITH-UHFFFAOYSA-N
SMILES: CC1=NN(C(=O)C1)C1=CC=CC(=C1)S(N)(=O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results