accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOOHNO

CAS RN 89-40-7

IUPAC Name: 5-nitro-2,3-dihydro-1H-isoindole-1,3-dione
Synonyms: 4-nitrophthalimide
Molecular Weight (g/mol): 192.13
Molecular Formula: C8H4N2O4
InChi Key: ANYWGXDASKQYAD-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C2C(=O)NC(=O)C2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results