accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONOOOHO

CAS RN 89-41-8

IUPAC Name: 4-methoxy-3-nitrobenzoic acid
Synonyms: 4-methoxy-3-nitrobenzoic acid
Molecular Weight (g/mol): 197.15
Molecular Formula: C8H7NO5
InChi Key: ANXBDAFDZSXOPQ-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1[N+]([O-])=O)C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)