accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 89-51-0

IUPAC Name: 2-(carboxymethyl)benzoic acid
Synonyms: homophthalic acid
Molecular Weight (g/mol): 180.16
Molecular Formula: C9H8O4
InChi Key: ZHQLTKAVLJKSKR-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC=CC=C1C(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (4)
Percent Purity 
  • (1)
  • (4)
  • (2)
Quantity 
  • (2)
  • (3)
  • (1)
  • (1)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (1)
  • (4)
  • (3)
  • (4)
  • (1)
  • (4)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)