accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ClNOO

CAS RN 89-59-8

IUPAC Name: 4-chloro-1-methyl-2-nitrobenzene
Synonyms: 4-chloro-1-methyl-2-nitrobenzene
Molecular Weight (g/mol): 171.58
Molecular Formula: C7H6ClNO2
InChi Key: SQFLFRQWPBEDHM-UHFFFAOYSA-N
SMILES: CC1=CC=C(Cl)C=C1[N+]([O-])=O
Boiling Point 
  • (1)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)