accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 89-63-4

IUPAC Name: 4-chloro-2-nitroaniline
Synonyms: 4-chloro-2-nitroaniline
Molecular Weight (g/mol): 172.57
Molecular Formula: C6H5ClN2O2
InChi Key: PBGKNXWGYQPUJK-UHFFFAOYSA-N
SMILES: NC1=CC=C(Cl)C=C1[N+]([O-])=O
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)