accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOOHHO

CAS RN 89-86-1

IUPAC Name: 2,4-dihydroxybenzoic acid
Synonyms: 2,4-dihydroxybenzoic acid resorcylic acid, β BRA β resorcylic acid β-resorcylic acid β-resorcinolic acid
Molecular Weight (g/mol): 154.12
Molecular Formula: C7H6O4
InChi Key: UIAFKZKHHVMJGS-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(O)C=C1O
Molecular Weight (g/mol) 
  • (5)
  • (4)
Percent Purity 
  • (1)
  • (8)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (1)
  • (6)
  • (5)
  • (4)
  • (1)
  • (8)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (4)