accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOH3COOCH3

CAS RN 89-91-8

IUPAC Name: methyl 2,2-dimethoxyacetate
Synonyms: methyl 2,2-dimethoxyacetate
Molecular Weight (g/mol): 134.13
Molecular Formula: C5H10O4
InChi Key: NZTCVGHPDWAALP-UHFFFAOYSA-N
SMILES: COC(OC)C(=O)OC
Boiling Point 
  • (5)
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (3)
  • (5)
Percent Purity 
  • (2)
  • (3)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (5)
  • (5)
  • (2)
  • (1)
  • (3)
  • (5)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)