accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HO

CAS RN 89-95-2

IUPAC Name: (2-methylphenyl)methanol
Synonyms: 2-methylbenzyl alcohol
Molecular Weight (g/mol): 122.17
Molecular Formula: C8H10O
InChi Key: XPNGNIFUDRPBFJ-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1CO
Boiling Point 
  • (1)
  • (4)
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (1)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (4)
  • (1)
  • (4)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)