accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NOON

CAS RN 89001-53-6

IUPAC Name: 2-methyl-4-nitrobenzonitrile
Synonyms: 2-methyl-4-nitrobenzonitrile
Molecular Weight (g/mol): 162.15
Molecular Formula: C8H6N2O2
InChi Key: RNTFKDBRMXYEPR-UHFFFAOYSA-N
SMILES: CC1=C(C=CC(=C1)[N+]([O-])=O)C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results