CAS RN 890148-78-4
IUPAC Name:
tris(3-bromophenyl)-1,3,5-triazine
Synonyms:
tris(3-bromophenyl)-1,3,5-triazine
Molecular Weight (g/mol):
546.06
Molecular Formula:
C21H12Br3N3
InChi Key:
IWHHYACTSSPXDV-UHFFFAOYSA-N
SMILES:
BrC1=CC=CC(=C1)C1=NC(=NC(=N1)C1=CC(Br)=CC=C1)C1=CC(Br)=CC=C1
1
–
1
of
1
results
1
2,4,6-Tris(3-bromophenyl)-1,3,5-triazine 98.0+%, TCI America™
CAS: 890148-78-4 Molecular Formula: C21H12Br3N3 Molecular Weight (g/mol): 546.06 InChI Key: IWHHYACTSSPXDV-UHFFFAOYSA-N PubChem CID: 57872377 IUPAC Name: 2,4,6-tris(3-bromophenyl)-1,3,5-triazine SMILES: C1=CC(=CC(=C1)Br)C2=NC(=NC(=N2)C3=CC(=CC=C3)Br)C4=CC(=CC=C4)Br