accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONOOCH3CH3

CAS RN 89028-40-0

IUPAC Name: 4-(propan-2-yl)-3-propanoyl-1,3-oxazolidin-2-one
Synonyms: 4-isopropyl-3-propanoyl-1,3-oxazolidin-2-one
Molecular Weight (g/mol): 185.22
Molecular Formula: C9H15NO3
InChi Key: HOWPHXVPNNPSAZ-UHFFFAOYNA-N
SMILES: CCC(=O)N1C(COC1=O)C(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results