accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 89044-79-1

IUPAC Name: 7-phenyl-1,4-diazepan-5-one
Synonyms: 7-phenyl-1,4-diazepan-5-one
Molecular Weight (g/mol): 190.25
Molecular Formula: C11H14N2O
InChi Key: UATUVKKDKVFGGB-UHFFFAOYNA-N
SMILES: O=C1CC(NCCN1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)