accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 890652-03-6

IUPAC Name: 1-(4-fluorophenyl)-1H-pyrazole-4-carbaldehyde
Synonyms: 1-(4-fluorophenyl)pyrazole-4-carbaldehyde
Molecular Weight (g/mol): 190.18
Molecular Formula: C10H7FN2O
InChi Key: OTZFOHVFXOKYGB-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1)N1C=C(C=O)C=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results