accessibility menu, dialog, popup

UserName

Viewing profile as user:
SOONHNH2HCl

CAS RN 89108-46-3

IUPAC Name: N-(4-aminobutyl)naphthalene-2-sulfonamide hydrochloride
Synonyms: N-(4-aminobutyl)naphthalene-2-sulfonamide hydrochloride
Molecular Weight (g/mol): 314.83
Molecular Formula: C14H19ClN2O2S
InChi Key: WYFVKUWCEVMLOL-UHFFFAOYSA-N
SMILES: Cl.NCCCCNS(=O)(=O)C1=CC2=CC=CC=C2C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)