accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OHNCH3H2N

CAS RN 89226-12-0

IUPAC Name: 2-amino-N,3,3-trimethylbutanamide
Synonyms: 2-amino-N,3,3-trimethylbutanamide
Molecular Weight (g/mol): 144.22
Molecular Formula: C7H16N2O
InChi Key: BPKJNEIOHOEWLO-UHFFFAOYNA-N
SMILES: CNC(=O)C(N)C(C)(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results