accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOHNClClCl

CAS RN 89238-99-3

IUPAC Name: (4-methoxyphenyl)methyl 2,2,2-trichloroethanimidate
Synonyms: (4-methoxyphenyl)methyl 2,2,2-trichloroethanimidate
Molecular Weight (g/mol): 282.55
Molecular Formula: C10H10Cl3NO2
InChi Key: TYHGKLBJBHACOI-UHFFFAOYSA-N
SMILES: COC1=CC=C(COC(=N)C(Cl)(Cl)Cl)C=C1
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)