accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NONBr

CAS RN 892501-91-6

IUPAC Name: 5-[2-(bromomethyl)phenyl]-3-methyl-1,2,4-oxadiazole
Synonyms: 5-[2-(bromomethyl)phenyl]-3-methyl-1,2,4-oxadiazole
Molecular Weight (g/mol): 253.1
Molecular Formula: C10H9BrN2O
InChi Key: GTXIOLDSSMKRDR-UHFFFAOYSA-N
SMILES: CC1=NOC(=N1)C1=CC=CC=C1CBr

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results