accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNFFF

CAS RN 892501-99-4

IUPAC Name: 4-[5-(trifluoromethyl)pyridin-2-yl]benzonitrile
Synonyms: 4-[5-(trifluoromethyl)pyridin-2-yl]benzonitrile
Molecular Weight (g/mol): 248.21
Molecular Formula: C13H7F3N2
InChi Key: DFPKSXZRVOQLJC-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CN=C(C=C1)C1=CC=C(C=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results