accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNFFFHO

CAS RN 892502-29-3

IUPAC Name: {2-[1-methyl-3-(trifluoromethyl)-1H-pyrazol-5-yl]phenyl}methanol
Synonyms: {2-[2-methyl-5-(trifluoromethyl)pyrazol-3-yl]phenyl}methanol
Molecular Weight (g/mol): 256.23
Molecular Formula: C12H11F3N2O
InChi Key: HLMRFDCLSUXVQC-UHFFFAOYSA-N
SMILES: CN1N=C(C=C1C1=CC=CC=C1CO)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results