accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 89282-12-2

IUPAC Name: 4-hydroxy-3-nitro-1,2-dihydropyridin-2-one
Synonyms: 4-hydroxy-3-nitro-1H-pyridin-2-one
Molecular Weight (g/mol): 156.1
Molecular Formula: C5H4N2O4
InChi Key: BKYGVGWYPFVKTK-UHFFFAOYSA-N
SMILES: OC1=C(C(=O)NC=C1)[N+]([O-])=O
Grade 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results