accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 893744-01-9

IUPAC Name: 5-[(morpholin-4-yl)methyl]thiophene-2-carbaldehyde
Synonyms: 5-(morpholin-4-ylmethyl)thiophene-2-carbaldehyde
Molecular Weight (g/mol): 211.28
Molecular Formula: C10H13NO2S
InChi Key: YPXGCMYNDMFOHE-UHFFFAOYSA-N
SMILES: O=CC1=CC=C(CN2CCOCC2)S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results