accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNHONOO

CAS RN 89399-21-3

IUPAC Name: N-hexyl-4-nitrobenzamide
Synonyms: N-hexyl-4-nitrobenzamide
Molecular Weight (g/mol): 250.3
Molecular Formula: C13H18N2O3
InChi Key: YIQCYVHPIUGJKJ-UHFFFAOYSA-N
SMILES: CCCCCCNC(=O)C1=CC=C(C=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results