accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSNNHOSNN

CAS RN 893990-34-6

IUPAC Name: N-(4-methyl-1,3-thiazol-2-yl)-2-[(6-phenylpyridazin-3-yl)sulfanyl]acetamide
Synonyms: N-(4-methyl-1,3-thiazol-2-yl)-2-[(6-phenylpyridazin-3-yl)sulfanyl]acetamide
Molecular Weight (g/mol): 342.44
Molecular Formula: C16H14N4OS2
InChi Key: WJRWSLORVIHRNX-UHFFFAOYSA-N
SMILES: CC1=CSC(NC(=O)CSC2=NN=C(C=C2)C2=CC=CC=C2)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results