CAS RN 894406-63-4
CAS RN 894406-63-4
IUPAC Name:
2-(trifluoromethyl)-[1,3]oxazolo[4,5-b]pyridine
Synonyms:
2-(trifluoromethyl)-[1,3]oxazolo[4,5-b]pyridine
Molecular Weight (g/mol):
188.11
Molecular Formula:
C7H3F3N2O
InChi Key:
MJILPMZYOJELAX-UHFFFAOYSA-N
SMILES:
FC(F)(F)C1=NC2=NC=CC=C2O1
1
–
1
of
1
results
1
Thermo Scientific Chemicals 2-(Trifluoromethyl)oxazolo[4,5-b]pyridine, 97%, Thermo Scientific™
CAS: 894406-63-4 Molecular Formula: C7H3F3N2O Molecular Weight (g/mol): 188.109 InChI Key: MJILPMZYOJELAX-UHFFFAOYSA-N PubChem CID: 29919200 IUPAC Name: 2-(trifluoromethyl)-[1,3]oxazolo[4,5-b]pyridine SMILES: C1=CC2=C(N=C1)N=C(O2)C(F)(F)F