accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NOOBOHHO

CAS RN 89466-07-9

IUPAC Name: (4-amino-3-nitrophenyl)boronic acid
Synonyms: 4-amino-3-nitrophenylboronic acid
Molecular Weight (g/mol): 181.94
Molecular Formula: C6H7BN2O4
InChi Key: IRTXQNNJQZNKRP-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1[N+]([O-])=O)B(O)O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results