accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NOO

CAS RN 89479-66-3

IUPAC Name: 3-methyl-5-phenyl-1,2-oxazole-4-carbaldehyde
Synonyms: 3-methyl-5-phenyl-1,2-oxazole-4-carbaldehyde
Molecular Weight (g/mol): 187.2
Molecular Formula: C11H9NO2
InChi Key: IZULDLZFUWWFHF-UHFFFAOYSA-N
SMILES: CC1=NOC(=C1C=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results