accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 89488-30-2

IUPAC Name: 5-bromo-3-methyl-1,2-dihydropyridin-2-one
Synonyms: 5-bromo-3-methyl-1H-pyridin-2-one
Molecular Weight (g/mol): 188.02
Molecular Formula: C6H6BrNO
InChi Key: HXPMRPRBABWPKL-UHFFFAOYSA-N
SMILES: CC1=CC(Br)=CNC1=O
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)