accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 89501-90-6

IUPAC Name: 1-methyl-1H-pyrazole-3-sulfonyl chloride
Synonyms: 1-methylpyrazole-3-sulfonyl chloride
Molecular Weight (g/mol): 180.61
Molecular Formula: C4H5ClN2O2S
InChi Key: TWLAHGNFQBQYEL-UHFFFAOYSA-N
SMILES: CN1C=CC(=N1)S(Cl)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results