accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COO(S)HNOOCH3H3CCH3POOCH3OCH3

CAS RN 89524-98-1

IUPAC Name: methyl (2S)-2-{[(tert-butoxy)carbonyl]amino}-2-(dimethoxyphosphoryl)acetate
Synonyms: methyl (2S)-2-[(tert-butoxycarbonyl)amino]-2-(dimethoxyphosphoryl)acetate
Molecular Weight (g/mol): 297.24
Molecular Formula: C10H20NO7P
InChi Key: LJHAPRKTPAREGO-ZETCQYMHSA-N
SMILES: COC(=O)[C@@H](NC(=O)OC(C)(C)C)P(=O)(OC)OC
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (3)
  • (1)
  • (3)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (3)
  • (2)
  • (2)