CAS RN 89547-15-9
IUPAC Name:
2-hexadecyloctadecanoic acid
Synonyms:
2-hexadecyloctadecanoic acid
Molecular Weight (g/mol):
508.92
Molecular Formula:
C34H68O2
InChi Key:
LPZKXVVBAZTEMK-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCCCCCCC(CCCCCCCCCCCCCCCC)C(O)=O
1
–
1
of
1
results
1
2-Hexadecyloctadecanoic Acid 98.0+%, TCI America™
CAS: 89547-15-9 Molecular Formula: C34H68O2 Molecular Weight (g/mol): 508.916 MDL Number: MFCD00191519 InChI Key: LPZKXVVBAZTEMK-UHFFFAOYSA-N Synonym: 2-Hexadecylstearic Acid PubChem CID: 625165 IUPAC Name: 2-hexadecyloctadecanoic acid SMILES: CCCCCCCCCCCCCCCCC(CCCCCCCCCCCCCCCC)C(=O)O