accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 89599-01-9

IUPAC Name: 3-bromobenzene-1-sulfonamide
Synonyms: benzenesulfonamide, 3-bromo-
Molecular Weight (g/mol): 236.08
Molecular Formula: C6H6BrNO2S
InChi Key: MUBJNMWVQGHHLG-UHFFFAOYSA-N
SMILES: NS(=O)(=O)C1=CC=CC(Br)=C1
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results