accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3NNOO(E/Z)

CAS RN 896-05-9

IUPAC Name: N,N-dimethyl-4-{[(4-nitrophenyl)imino]methyl}aniline
Synonyms: N,N-dimethyl-4-{[(4-nitrophenyl)imino]methyl}aniline
Molecular Weight (g/mol): 269.3
Molecular Formula: C15H15N3O2
InChi Key: RLDPRVKBHPQKIK-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=C(C=NC2=CC=C(C=C2)[N+]([O-])=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results