CAS RN 89601-24-1
IUPAC Name:
1-octadecyl-1H-pyrrole
Synonyms:
1-octadecylpyrrole
Molecular Weight (g/mol):
319.58
Molecular Formula:
C22H41N
InChi Key:
BMSMIJNAHWEOBQ-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCCCCCCCCN1C=CC=C1
1
–
1
of
1
results
1
1-Octadecylpyrrole 95.0+%, TCI America™
CAS: 89601-24-1 Molecular Formula: C22H41N Molecular Weight (g/mol): 319.577 MDL Number: MFCD00144990 InChI Key: BMSMIJNAHWEOBQ-UHFFFAOYSA-N PubChem CID: 19013669 IUPAC Name: 1-octadecylpyrrole SMILES: CCCCCCCCCCCCCCCCCCN1C=CC=C1