accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 89686-69-1

IUPAC Name: (2S)-2-{2-[(2S)-oxolan-2-yl]propan-2-yl}oxolane
Synonyms: ditetrahydrofurylpropane
Molecular Weight (g/mol): 184.28
Molecular Formula: C11H20O2
InChi Key: FZLHAQMQWDDWFI-UWVGGRQHSA-N
SMILES: CC(C)([C@@H]1CCCO1)[C@@H]1CCCO1
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)